siRNA Details
| Gene Symbol: C10orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024942.3 | NP_079218.2 | 2 | 548 | ||
| GenBank mRNA | AK023552.1 | - | 2 | 404 | ||
| BC032645.1 | - | 2 | 388 | 5519534 | ||
| BI821345.1 | - | 2 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf88| Gene Symbol: C10orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024942.3 | NP_079218.2 | 2 | 548 | ||
| GenBank mRNA | AK023552.1 | - | 2 | 404 | ||
| BC032645.1 | - | 2 | 388 | 5519534 | ||
| BI821345.1 | - | 2 | 548 | |||