siRNA Details
| Gene Symbol: FAM188A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318330.1 | NP_001305259.1 | 13 | 1349 | ||
| NM_024948.3 | NP_079224.1 | 14 | 1401 | |||
| XM_005252600.2 | XP_005252657.1 | 13 | 1295 | |||
| XM_005252602.2 | XP_005252659.1 | 12 | 1243 | |||
| XM_006717508.2 | XP_006717571.1 | 13 | 1297 | |||
| XM_011519690.2 | XP_011517992.1 | 14 | 1378 | |||
| XM_011519693.1 | XP_011517995.1 | 14 | 1387 | |||
| XM_011519694.1 | XP_011517996.1 | 15 | 1442 | |||
| XM_017016671.1 | XP_016872160.1 | 13 | 1297 | |||
| XM_017016672.1 | XP_016872161.1 | 16 | 1534 | |||
| XM_017016673.1 | XP_016872162.1 | 14 | 1404 | |||
| XM_017016674.1 | XP_016872163.1 | 12 | 1245 | |||
| XM_017016675.1 | XP_016872164.1 | 12 | 1214 | |||
| XM_017016676.1 | XP_016872165.1 | 11 | 1162 | |||
| GenBank mRNA | AB014761.1 | - | 14 | 1167 | ||
| AF063600.1 | - | 9 | 667 | |||
| AF176916.1 | - | 13 | 1311 | |||
| AK023459.1 | - | 14 | 1388 | |||
| AL833873.1 | - | 14 | 1363 | |||
| BC020605.1 | - | 13 | 1314 | |||
| BC067799.1 | - | 14 | 1387 | 5278533 | ||
| CR457315.1 | - | 14 | 1167 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024185, BC002225, BC027202, AK009891, AK049869, and others.

FAM188A
Genomic Map