siRNA Details
| Gene Symbol: ZNF606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025027.3 | NP_079303.2 | 7 | 1945 | ||
| XM_005259276.1 | XP_005259333.1 | 7 | 1805 | |||
| GenBank mRNA | AB058755.1 | - | 7 | 1948 | ||
| AF455357.1 | - | 8 | 2507 | |||
| AK293097.1 | - | 7 | 1415 | |||
| AK294037.1 | - | 6 | 1329 | |||
| BC037209.1 | - | 7 | 1419 | 4837545 | ||

ZNF606
Genomic Map