siRNA Details
| Gene Symbol: ZNF614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025040.3 | NP_079316.2 | 5 | 2105 | ||
| GenBank mRNA | AK025594.1 | - | 1 | 507 | ||
| AK097156.1 | - | 5 | 2089 | |||
| BC022246.1 | - | 1 | 904 | |||
| BC101392.1 | - | 5 | 1943 | 40026900 | ||
| BC101393.2 | - | 5 | 1943 | 40026901 | ||
| BC101394.1 | - | 5 | 1943 | 40026907 | ||
| BC101395.1 | - | 5 | 1943 | 40026909 | ||

ZNF614
Genomic Map