siRNA Details
| Gene Symbol: BBOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025057.2 | NP_079333.2 | 8 | 1375 | ||
| XM_005268092.3 | XP_005268149.1 | 8 | 1381 | |||
| XM_005268098.4 | XP_005268155.1 | 6 | 1014 | |||
| XM_011537170.2 | XP_011535472.1 | 9 | 1489 | |||
| XM_011537171.2 | XP_011535473.1 | 9 | 1489 | |||
| XM_011537172.1 | XP_011535474.1 | 9 | 1489 | |||
| XM_011537174.2 | XP_011535476.1 | 9 | 1489 | |||
| XM_011537175.2 | XP_011535477.1 | 8 | 1345 | |||
| XM_011537176.2 | XP_011535478.1 | 10 | 1473 | |||
| XM_011537177.2 | XP_011535479.1 | 8 | 1193 | |||
| XM_011537178.2 | XP_011535480.1 | 8 | 1164 | |||
| XM_011537179.2 | XP_011535481.1 | 7 | 1049 | |||
| XM_017021659.1 | XP_016877148.1 | 8 | 1381 | |||
| XM_017021660.1 | XP_016877149.1 | 7 | 1097 | |||
| XM_017021661.1 | XP_016877150.1 | 7 | 1149 | |||
| XM_017021662.1 | XP_016877151.1 | 6 | 933 | |||
| GenBank mRNA | AK026842.1 | - | 1 | 408 | ||
| AK097531.1 | - | 6 | 1053 | |||
| AL834487.1 | - | 3 | 650 | |||
| BC104979.1 | - | 1 | 406 | |||
| BC104981.1 | - | 1 | 406 | |||

BBOF1
Genomic Map