siRNA Details
| Gene Symbol: CCDC170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025059.3 | NP_079335.2 | 11 | 2097 | ||
| XM_011536147.2 | XP_011534449.1 | 11 | 2135 | |||
| XM_011536148.2 | XP_011534450.1 | 10 | 1937 | |||
| GenBank mRNA | AK026958.1 | - | 11 | 2010 | ||
| AK027300.1 | - | 3 | 504 | |||
| AK225330.1 | - | 11 | 2011 | |||
| BC060803.1 | - | 11 | 2046 | 5298914 | ||

CCDC170
Genomic Map