siRNA Details
| Gene Symbol: CCDC170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025059.3 | NP_079335.2 | 5 | 723 | ||
| XM_011536147.2 | XP_011534449.1 | 5 | 761 | |||
| XM_011536148.2 | XP_011534450.1 | 5 | 764 | |||
| GenBank mRNA | AK026958.1 | - | 5 | 636 | ||
| AK225330.1 | - | 5 | 637 | |||
| BC060803.1 | - | 5 | 672 | 5298914 | ||

CCDC170
Genomic Map