siRNA Details
| Gene Symbol: MROH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163629.1 | NP_001157101.1 | 9 | 859 | ||
| NM_025063.2 | NP_079339.2 | 9 | 835 | |||
| XM_011510005.2 | XP_011508307.1 | 9 | 859 | |||
| XM_011510006.2 | XP_011508308.1 | 9 | 859 | |||
| GenBank mRNA | AK027203.1 | - | 9 | 803 | ||
| BC113390.1 | - | 9 | 790 | |||
| BC113392.1 | - | 9 | 790 | |||

MROH9
Genomic Map