siRNA Details
| Gene Symbol: ZNF703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025069.2 | NP_079345.1 | 2 | 1959 | ||
| GenBank mRNA | AK024361.1 | - | 2 | 1927 | ||
| BC032534.1 | - | 2 | 1888 | 5527569 | ||
| BC084581.1 | - | 3 | 640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF703| Gene Symbol: ZNF703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025069.2 | NP_079345.1 | 2 | 1959 | ||
| GenBank mRNA | AK024361.1 | - | 2 | 1927 | ||
| BC032534.1 | - | 2 | 1888 | 5527569 | ||
| BC084581.1 | - | 3 | 640 | |||