siRNA Details
| Gene Symbol: FRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166133.1 | NP_001159605.1 | 6, 7 | 1051 | ||
| NM_025074.6 | NP_079350.5 | 6, 7 | 1051 | |||
| XM_006714316.2 | XP_006714379.1 | 6, 7 | 1298 | |||
| GenBank mRNA | AK027833.1 | - | 1, 2 | 36 | ||
| BC052281.1 | - | 1, 2 | 138 | |||
| BC064487.1 | - | 1, 2 | 138 | |||
| BC131820.1 | - | 6, 7 | 641 | |||
| BC131821.1 | - | 6, 7 | 641 | |||

FRAS1
Genomic Map