siRNA Details
| Gene Symbol: CTBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004388.2 | NP_004379.1 | 4 | 749 | ||
| GenBank mRNA | AK075423.1 | - | 4 | 1013 | ||
| AK298703.1 | - | 4 | 700 | |||
| AK314640.1 | - | 4 | 715 | |||
| BC024007.2 | - | 3 | 531 | |||
| BC096752.1 | - | 3 | 531 | |||
| BC126333.1 | - | 4 | 725 | 8992049 | ||
| BC126335.1 | - | 4 | 725 | 8992051 | ||
| M95767.1 | - | 4 | 684 | |||

CTBS
Genomic Map