siRNA Details
| Gene Symbol: CWH43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286791.1 | NP_001273720.1 | 9 | 1328 | ||
| NM_025087.2 | NP_079363.2 | 9 | 1409 | |||
| XM_011513755.1 | XP_011512057.1 | 9 | 1410 | |||
| XM_011513756.2 | XP_011512058.1 | 9 | 1441 | |||
| XM_011513757.2 | XP_011512059.1 | 9 | 1467 | |||
| XM_011513758.1 | XP_011512060.1 | 8 | 1152 | |||
| XM_011513759.1 | XP_011512061.1 | 6 | 806 | |||
| GenBank mRNA | AK025164.1 | - | 9 | 1409 | ||
| AK300495.1 | - | 9 | 1331 | |||
| AK316307.1 | - | 9 | 1301 | |||
| BC137387.1 | - | 9 | 1392 | 9021385 | ||
| BC137388.1 | - | 9 | 1392 | 9021386 | ||

CWH43
Genomic Map