siRNA Details
| Gene Symbol: C16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025108.2 | NP_079384.2 | 4 | 354 | ||
| XM_011522667.1 | XP_011520969.1 | 3 | 258 | |||
| GenBank mRNA | AK023971.1 | - | 3 | 435 | ||
| AK093788.1 | - | 3 | 592 | |||
| AK301929.1 | - | 4 | 350 | |||
| AL559561.3 | - | 4 | 354 | |||
| BC018719.1 | - | 4 | 342 | 4868952 | ||

C16orf59
Genomic Map