siRNA Details
| Gene Symbol: MYO19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033580.2 | NP_001028752.1 | 8 | 1553 | ||
| NM_001163735.1 | NP_001157207.1 | 8 | 1553 | |||
| NM_025109.5 | NP_079385.2 | 8 | 1553 | |||
| XM_011525286.1 | XP_011523588.1 | 9 | 1102 | |||
| XM_011525287.1 | XP_011523589.1 | 9 | 1102 | |||
| XM_011525290.2 | XP_011523592.1 | 9 | 1102 | |||
| XM_017025157.1 | XP_016880646.1 | 9 | 1076 | |||
| XM_017025158.1 | XP_016880647.1 | 8 | 1009 | |||
| XM_017025159.1 | XP_016880648.1 | 9 | 1102 | |||
| XM_017025160.1 | XP_016880649.1 | 9 | 1102 | |||
| GenBank mRNA | AB209035.1 | - | 8 | 1511 | ||
| AK026518.1 | - | 8 | 964 | |||
| AK299802.1 | - | 5 | 590 | |||
| AK304073.1 | - | 8 | 904 | |||
| BC008900.2 | - | 8 | 944 | 3629870 | ||

MYO19
Genomic Map