siRNA Details
| Gene Symbol: FHOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281739.2 | NP_001268668.1 | 3 | 438 | ||
| NM_001281740.2 | NP_001268669.1 | 3 | 438 | |||
| NM_025135.4 | NP_079411.2 | 3 | 438 | |||
| XM_005258349.1 | XP_005258406.1 | 3 | 420 | |||
| XM_005258354.1 | XP_005258411.1 | 3 | 420 | |||
| XM_005258355.1 | XP_005258412.1 | 3 | 420 | |||
| XM_011526189.1 | XP_011524491.1 | 3 | 420 | |||
| XM_011526190.1 | XP_011524492.1 | 3 | 420 | |||
| XM_011526192.1 | XP_011524494.1 | 3 | 420 | |||
| XM_011526193.2 | XP_011524495.1 | 3 | 420 | |||
| XM_011526195.1 | XP_011524497.1 | 3 | 420 | |||
| XM_011526196.1 | XP_011524498.1 | 3 | 420 | |||
| XM_011526197.1 | XP_011524499.1 | 3 | 420 | |||
| XM_017026006.1 | XP_016881495.1 | 3 | 420 | |||
| XM_017026008.1 | XP_016881497.1 | 3 | 420 | |||
| XM_017026009.1 | XP_016881498.1 | 3 | 420 | |||
| GenBank mRNA | AB084087.1 | - | 3 | 298 | ||
| AK128053.1 | - | 3 | 417 | |||
| HM191478.1 | - | 3 | 298 | |||

FHOD3
Genomic Map