siRNA Details
| Gene Symbol: PROSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025138.4 | NP_079414.3 | 5 | 1165 | ||
| XM_005266545.3 | XP_005266602.1 | 4 | 1138 | |||
| XM_011535239.2 | XP_011533541.1 | 5 | 1185 | |||
| GenBank mRNA | AB107354.1 | - | 5 | 1165 | ||
| AK074360.1 | - | 5 | 1138 | |||
| AK299663.1 | - | 2 | 161 | |||
| AL522407.3 | - | 4 | 864 | |||
| BC064348.1 | - | 5 | 420 | 4939198 | ||
| BX647471.1 | - | 5 | 587 | |||
| DA786460.1 | - | 2 | 161 | |||

PROSER1
Genomic Map