siRNA Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304957.1 | NP_001291886.1 | 3, 4 | 685 | ||
| NM_001304958.1 | NP_001291887.1 | 4, 5 | 792 | |||
| NM_001304959.1 | NP_001291888.1 | 3, 4 | 434 | |||
| NM_001304960.1 | NP_001291889.1 | 5, 6 | 603 | |||
| NM_001304961.1 | NP_001291890.1 | 2, 3 | 602 | |||
| NM_025140.2 | NP_079416.1 | 3, 4 | 695 | |||
| XM_005253624.2 | XP_005253681.1 | 3, 4 | 439 | |||
| XM_017019984.1 | XP_016875473.1 | 3, 4 | 1253 | |||
| GenBank mRNA | AB015292.1 | - | 2, 3 | 215 | ||
| AK026124.1 | - | 3, 4 | 326 | |||
| AK054680.1 | - | 1, 2 | 1962 | |||
| AK125866.1 | - | 1, 2 | 2213 | |||
| AK222661.1 | - | 3, 4 | 283 | |||
| AW953473.1 | - | 5, 6 | 574 | |||
| BC017186.2 | - | 3, 4 | 318 | 3051346 | ||
| BX394686.2 | - | 2, 3 | 351 | |||
| DA263704.1 | - | 3, 4 | 434 | |||
| DA370549.1 | - | 4, 5 | 390 | |||

CCDC92
Genomic Map