siRNA Details
| Gene Symbol: TM2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307960.1 | NP_001294889.1 | 4 | 494 | ||
| NM_001308026.1 | NP_001294955.1 | 5 | 572 | |||
| NM_025141.3 | NP_079417.2 | 4 | 494 | |||
| NM_078474.2 | NP_510883.2 | 5 | 572 | |||
| GenBank mRNA | AF353992.1 | - | 4 | 469 | ||
| AK026257.1 | - | 4 | 484 | |||
| AK094955.1 | - | 3 | 904 | |||
| AK297048.1 | - | 4 | 479 | |||
| AK315584.1 | - | 5 | 548 | |||
| AL512689.1 | - | 1 | 1330 | |||
| BC006150.1 | - | 5 | 544 | 3051281 | ||
| BC008873.2 | - | 4 | 467 | 4137334 | ||
| BU595412.1 | - | 5 | 567 | |||

TM2D3
Genomic Map