siRNA Details
| Gene Symbol: COQ10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320819.1 | NP_001307748.1 | 2 | 342 | ||
| NM_001320820.1 | NP_001307749.1 | 2 | 539 | |||
| NM_025147.4 | NP_079423.1 | 2 | 298 | |||
| GenBank mRNA | AK023510.1 | - | 2 | 167 | ||
| AK294093.1 | - | 2 | 539 | |||
| BC022453.1 | - | 2 | 215 | 4796819 | ||
| DC349756.1 | - | 2 | 298 | |||
| HY088880.1 | - | 2 | 291 | |||

COQ10B
Genomic Map