siRNA Details
| Gene Symbol: ACSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288968.1 | NP_001275897.1 | 13 | 1642 | ||
| NM_001288969.1 | NP_001275898.1 | 12 | 1528 | |||
| NM_001288970.1 | NP_001275899.1 | 11 | 1438 | |||
| NM_001288971.1 | NP_001275900.1 | 10 | 1242 | |||
| NM_001288972.1 | NP_001275901.1 | 12 | 1484 | |||
| NM_025149.5 | NP_079425.3 | 12 | 1567 | |||
| XM_011525294.1 | XP_011523596.1 | 12 | 1489 | |||
| GenBank mRNA | AK024573.1 | - | 12 | 1474 | ||
| AK225044.1 | - | 12 | 1474 | |||
| AK225561.1 | - | 12 | 1473 | |||
| AK294205.1 | - | 13 | 1584 | |||
| AK295258.1 | - | 12 | 1436 | |||
| AK300145.1 | - | 11 | 1346 | |||
| AK300625.1 | - | 10 | 1242 | |||
| AY358660.1 | - | 12 | 1503 | |||
| BC012053.1 | - | 12 | 1480 | 4577089 | ||
| BC014123.2 | - | 12 | 1487 | 4560029 | ||

ACSF2
Genomic Map