siRNA Details
| Gene Symbol: EFHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025184.3 | NP_079460.2 | 10 | 1674 | ||
| XM_006724562.2 | XP_006724625.1 | 9 | 1387 | |||
| GenBank mRNA | AK026496.1 | - | 10 | 1528 | ||
| AK291187.1 | - | 10 | 1815 | |||
| AK292967.1 | - | 10 | 1658 | |||
| BC031039.1 | - | 10 | 1669 | 5272398 | ||
| CR749363.1 | - | 5 | 1335 | |||

EFHC2
Genomic Map