siRNA Details
| Gene Symbol: EFHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025184.3 | NP_079460.2 | 2 | 310 | ||
| GenBank mRNA | AK026496.1 | - | 2 | 163 | ||
| AK291187.1 | - | 2 | 451 | |||
| AK292967.1 | - | 2 | 294 | |||
| AK297714.1 | - | 2 | 299 | |||
| BC031039.1 | - | 2 | 305 | 5272398 | ||
| DC331145.1 | - | 2 | 311 | |||

EFHC2
Genomic Map