siRNA Details
| Gene Symbol: C16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320540.1 | NP_001307469.1 | 11 | 968 | ||
| NM_001320541.1 | NP_001307470.1 | 11 | 1009 | |||
| NM_001320542.1 | NP_001307471.1 | 11 | 994 | |||
| NM_001320543.1 | NP_001307472.1 | 10 | 925 | |||
| NM_025187.4 | NP_079463.2 | 10 | 1024 | |||
| XM_006721284.3 | XP_006721347.1 | 10 | 1283 | |||
| XM_011523350.2 | XP_011521652.1 | 9 | 1423 | |||
| XM_017023729.1 | XP_016879218.1 | 10 | 1294 | |||
| GenBank mRNA | AF176088.1 | - | 11 | 924 | ||
| AL540304.3 | - | 11 | 903 | |||
| BC004556.2 | - | 10 | 1004 | 3957147 | ||

C16orf70
Genomic Map