siRNA Details
| Gene Symbol: ZNF430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172671.1 | NP_001166142.1 | 2 | 265 | ||
| NM_025189.3 | NP_079465.3 | 2 | 265 | |||
| GenBank mRNA | AK023721.1 | - | 2 | 245 | ||
| AK290475.1 | - | 2 | 265 | |||
| AY269787.1 | - | 2 | 194 | |||
| BC111776.1 | - | 2 | 153 | 40033720 | ||
| DB255551.1 | - | 2 | 228 | |||

ZNF430
Genomic Map