siRNA Details
| Gene Symbol: KLHL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030624.2 | NP_085127.2 | 4 | 1132 | ||
| XM_006724518.1 | XP_006724581.1 | 3 | 1344 | |||
| GenBank mRNA | AB051464.1 | - | 2 | 629 | ||
| AK057298.1 | - | 4 | 1132 | |||
| BC109058.2 | - | 2 | 876 | 40033682 | ||
| BC109059.2 | - | 2 | 876 | 40033683 | ||
| EU794630.1 | - | 2 | 876 | |||

KLHL15
Genomic Map