siRNA Details
| Gene Symbol: ZKSCAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242894.1 | NP_001229823.1 | 4, 5 | 875 | ||
| NM_001242895.1 | NP_001229824.1 | 2, 3 | 294 | |||
| NM_024493.3 | NP_077819.2 | 3, 4 | 758 | |||
| XM_005249423.3 | XP_005249480.1 | 2, 3 | 924 | |||
| XM_006715215.2 | XP_006715278.1 | 3, 4 | 656 | |||
| XM_006715216.2 | XP_006715279.1 | 3, 4 | 671 | |||
| XM_006715218.3 | XP_006715281.1 | 2, 3 | 222 | |||
| XM_017011320.1 | XP_016866809.1 | 4, 5 | 752 | |||
| GenBank mRNA | AK091225.1 | - | 2, 3 | 2092 | ||
| AK122790.1 | - | 2, 3 | 258 | |||
| AK313532.1 | - | 4, 5 | 839 | |||
| BC006118.1 | - | 3, 4 | 690 | 3505521 | ||
| BT007427.1 | - | 2, 3 | 542 | |||

ZKSCAN3
Genomic Map