siRNA Details
| Gene Symbol: PABPC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001124756.2 | NP_001118228.1 | 11 | 1541 | ||
| NR_134983.1 | 10 | 1392 | ||||
| NR_134984.1 | 2 | 1637 | ||||
| NR_134985.1 | 2 | 1637 | ||||
| NR_134986.1 | 2 | 1637 | ||||
| NR_134987.1 | 2 | 1637 | ||||
| GenBank mRNA | AK026760.1 | - | 2 | 1020 | ||
| AK124047.1 | - | 2 | 1637 | |||
| BC068104.1 | - | 2 | 821 | |||
| BM689138.1 | - | 3 | 167 | |||
| BM832739.1 | - | 2 | 98 | |||
| BX451955.2 | - | 4 | 497 | |||
| EU190483.1 | - | 10 | 1328 | |||

PABPC1L
Genomic Map