siRNA Details
| Gene Symbol: SLC19A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025243.3 | NP_079519.1 | 6 | 1464 | ||
| XM_005246874.3 | XP_005246931.1 | 7 | 1627 | |||
| XM_011511931.2 | XP_011510233.1 | 6 | 1719 | |||
| XM_011511932.1 | XP_011510234.1 | 6 | 1541 | |||
| XM_011511933.1 | XP_011510235.1 | 6 | 1807 | |||
| XM_017005030.1 | XP_016860519.1 | 7 | 1809 | |||
| XM_017005031.1 | XP_016860520.1 | 7 | 1788 | |||
| XM_017005032.1 | XP_016860521.1 | 7 | 1669 | |||
| XM_017005033.1 | XP_016860522.1 | 7 | 1983 | |||
| XM_017005034.1 | XP_016860523.1 | 7 | 1913 | |||
| GenBank mRNA | AF271633.1 | - | 6 | 1418 | ||
| AF283317.1 | - | 6 | 1445 | |||
| AK301490.1 | - | 7 | 1630 | |||
| AK312464.1 | - | 6 | 1466 | |||
| BC032014.1 | - | 6 | 1419 | 5177540 | ||

SLC19A3
Genomic Map