siRNA Details
| Gene Symbol: KIAA1683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145304.1 | NP_001138776.1 | 3 | 2125 | ||
| NM_001145305.1 | NP_001138777.1 | 3 | 2164 | |||
| NM_025249.3 | NP_079525.1 | 3 | 2125 | |||
| XM_005260084.1 | XP_005260141.1 | 3 | 2050 | |||
| XM_011528324.1 | XP_011526626.1 | 2 | 2071 | |||
| GenBank mRNA | AK093064.1 | - | 1 | 94 | ||
| AK302431.1 | - | 3 | 2077 | |||
| AL136867.1 | - | 3 | 2042 | |||
| BC112932.1 | - | 3 | 2051 | 5744104 | ||

KIAA1683
Genomic Map