siRNA Details
| Gene Symbol: TTYH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025250.2 | NP_079526.1 | 14 | 2548 | ||
| XM_017012656.1 | XP_016868145.1 | 13 | 2420 | |||
| GenBank mRNA | AB051478.2 | - | 14 | 2543 | ||
| AB162931.1 | - | 14 | 2373 | |||
| AF318350.1 | - | 4 | 1192 | |||
| AK074158.1 | - | 10 | 1856 | |||
| AK096851.1 | - | 2 | 892 | |||
| AK124608.1 | - | 10 | 1991 | |||
| AL833825.1 | - | 1 | 843 | |||
| BC131824.1 | - | 14 | 2419 | 40128913 | ||
| BC152447.1 | - | 14 | 2543 | 8862578 | ||

TTYH3
Genomic Map