siRNA Details
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316349.1 | NP_001303278.1 | 3 | 901 | ||
GenBank mRNA | AB051466.1 | - | 1 | 525 | ||
AK299929.1 | - | 4 | 1037 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316349.1 | NP_001303278.1 | 3 | 901 | ||
GenBank mRNA | AB051466.1 | - | 1 | 525 | ||
AK299929.1 | - | 4 | 1037 |