siRNA Details
| Gene Symbol: LY6G6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025261.2 | NP_079537.1 | 3 | 522 | ||
| GenBank mRNA | AY359119.1 | - | 3 | 522 | ||
| BC036302.1 | - | 3 | 497 | 4776231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY6G6C| Gene Symbol: LY6G6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025261.2 | NP_079537.1 | 3 | 522 | ||
| GenBank mRNA | AY359119.1 | - | 3 | 522 | ||
| BC036302.1 | - | 3 | 497 | 4776231 | ||