siRNA Details
| Gene Symbol: LY6G5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025262.3 | NP_079538.3 | 2 | 268 | ||
| GenBank mRNA | AJ315540.1 | - | 3 | 372 | ||
| AJ315541.1 | - | 3 | 492 | |||
| AJ315542.1 | - | 2 | 259 | |||
| AJ315543.1 | - | 2 | 353 | |||
| AJ315544.1 | - | 1 | 1243 | |||

LY6G5C
Genomic Map