siRNA Details
| Gene Symbol: AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261434.1 | NP_001248363.1 | 4 | 424 | ||
| GenBank mRNA | AK309477.1 | - | 4 | 371 | ||
| AL515977.3 | - | 4 | 369 | |||
| BP368900.1 | - | 4 | 402 | |||
| CA951145.1 | - | 4 | 393 | |||
| Gene Symbol: PTGES3L-AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136042.2 | NP_001129514.2 | 9 | 1234 | ||
| NM_025267.3 | NP_079543.1 | 9 | 850 | |||
| GenBank mRNA | AF039235.1 | - | 9 | 939 | ||
| AK094084.1 | - | 9 | 788 | |||
| AK295447.1 | - | 9 | 1234 | |||
| BC004172.2 | - | 9 | 850 | 2823004 | ||
| BC019324.1 | - | 9 | 850 | 2823004 | ||

AARSD1
Genomic Map