siRNA Details
| Gene Symbol: TMEM121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025268.2 | NP_079544.1 | 2 | 391 | ||
| XM_005268101.3 | XP_005268158.1 | 2 | 998 | |||
| XM_006720261.3 | XP_006720324.1 | 2 | 830 | |||
| GenBank mRNA | BC004221.2 | - | 2 | 391 | 3530864 | |

TMEM121
Genomic Map