siRNA Details
| Gene Symbol: THAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008695.1 | NP_001008695.1 | 3, 4 | 267 | ||
| NM_030573.2 | NP_085050.2 | 2, 3 | 441 | |||
| GenBank mRNA | AK315457.1 | - | 2, 3 | 426 | ||
| BC004346.1 | - | 2, 3 | 412 | 3633743 | ||
| BE264280.1 | - | 2, 3 | 340 | |||
| BI223500.1 | - | 2, 3 | 384 | |||
| BM790105.1 | - | 3, 4 | 267 | |||
| CR456343.1 | - | 2, 3 | 259 | |||

THAP7
Genomic Map