siRNA Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030576.3 | NP_085053.1 | 5 | 1290 | ||
| XM_005257703.1 | XP_005257760.1 | 5 | 1221 | |||
| XM_005257705.3 | XP_005257762.1 | 5 | 1401 | |||
| XM_006722124.1 | XP_006722187.1 | 5 | 1197 | |||
| GenBank mRNA | AK092301.1 | - | 5 | 1256 | ||
| BC004400.1 | - | 5 | 1271 | 3636469 | ||

LIMD2
Genomic Map