siRNA Details
| Gene Symbol: TMEM177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105198.1 | NP_001098668.1 | 2 | 1326 | ||
| NM_001105199.1 | NP_001098669.1 | 2 | 934 | |||
| NM_030577.2 | NP_085054.2 | 2 | 985 | |||
| GenBank mRNA | AK096890.1 | - | 2 | 931 | ||
| AK130088.1 | - | 2 | 934 | |||
| BC004404.1 | - | 2 | 973 | 3637432 | ||

TMEM177
Genomic Map