siRNA Details
| Gene Symbol: TMEM177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105198.1 | NP_001098668.1 | 2 | 691 | ||
| NM_001105199.1 | NP_001098669.1 | 2 | 299 | |||
| NM_030577.2 | NP_085054.2 | 2 | 350 | |||
| GenBank mRNA | AK096890.1 | - | 2 | 296 | ||
| AK130088.1 | - | 2 | 299 | |||
| AK299078.1 | - | 2 | 296 | |||
| AK308503.1 | - | 2 | 314 | |||
| BC004404.1 | - | 2 | 338 | 3637432 | ||
| BG280893.1 | - | 2 | 293 | |||
| BG776153.1 | - | 2 | 321 | |||

TMEM177
Genomic Map