siRNA Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317237.1 | NP_001304166.1 | 4, 5 | 480 | ||
| NM_001908.4 | NP_001899.1 | 3, 4 | 392 | |||
| NM_147780.3 | NP_680090.1 | 5, 6 | 554 | |||
| NM_147781.3 | NP_680091.1 | 4, 5 | 511 | |||
| NM_147782.3 | NP_680092.1 | 4, 5 | 480 | |||
| NM_147783.3 | NP_680093.1 | 4, 5 | 466 | |||
| XM_006716244.2 | XP_006716307.1 | 5, 6 | 578 | |||
| XM_006716245.2 | XP_006716308.1 | 3, 4 | 682 | |||
| XM_011543812.2 | XP_011542114.1 | 5, 6 | 487 | |||
| XM_017013097.1 | XP_016868586.1 | 4, 5 | 468 | |||
| XM_017013098.1 | XP_016868587.1 | 4, 5 | 468 | |||
| XM_017013099.1 | XP_016868588.1 | 5, 6 | 510 | |||
| XM_017013100.1 | XP_016868589.1 | 4, 5 | 412 | |||
| GenBank mRNA | AK075393.1 | - | 3, 4 | 368 | ||
| AK092070.1 | - | 4, 5 | 403 | |||
| AK130184.1 | - | 3, 4 | 329 | |||
| AK290239.1 | - | 3, 4 | 329 | |||
| AK296845.1 | - | 3, 4 | 331 | |||
| AL543654.3 | - | 4, 5 | 440 | |||
| BC010240.1 | - | 4, 5 | 409 | 4154307 | ||
| BC095408.1 | - | 3, 4 | 329 | 30334082 | ||
| BG770805.1 | - | 5, 6 | 463 | |||
| BM008741.1 | - | 3, 4 | 368 | |||
| BX647765.1 | - | 4, 5 | 371 | |||
| BX647952.1 | - | 3, 4 | 332 | |||
| CB997355.1 | - | 3, 4 | 406 | |||
| DC386022.1 | - | 3, 4 | 392 | |||
| L16510.1 | - | 4, 5 | 394 | |||
| M14221.1 | - | 4, 5 | 411 | |||

CTSB
Genomic Map