siRNA Details
| Gene Symbol: ASXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030632.2 | NP_085135.1 | 12 | 3218 | ||
| XM_005258356.1 | XP_005258413.1 | 13 | 3191 | |||
| XM_011526205.2 | XP_011524507.1 | 13 | 9981 | |||
| XM_011526206.2 | XP_011524508.1 | 13 | 6746 | |||
| XM_011526209.1 | XP_011524511.1 | 14 | 3375 | |||
| XM_011526210.1 | XP_011524512.1 | 14 | 3432 | |||
| XM_011526212.1 | XP_011524514.1 | 13 | 3336 | |||
| XM_011526213.1 | XP_011524515.1 | 13 | 3429 | |||
| XM_017026012.1 | XP_016881501.1 | 13 | 6593 | |||
| XM_017026013.1 | XP_016881502.1 | 14 | 6955 | |||
| XM_017026014.1 | XP_016881503.1 | 14 | 3392 | |||
| GenBank mRNA | AB051500.2 | - | 2 | 1377 | ||
| AK131454.1 | - | 14 | 3261 | |||
| AL133050.1 | - | 2 | 713 | |||

ASXL3
Genomic Map