siRNA Details
| Gene Symbol: CEP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040157.2 | NP_001035247.1 | 7, 8 | 1100 | ||
| NM_001145314.1 | NP_001138786.1 | 7, 8 | 991 | |||
| XM_011532285.2 | XP_011530587.1 | 7, 8 | 1051 | |||
| XM_011532286.2 | XP_011530588.1 | 7, 8 | 1100 | |||
| XM_011532287.2 | XP_011530589.1 | 6, 7 | 950 | |||
| XM_017008654.1 | XP_016864143.1 | 7, 8 | 1051 | |||
| XM_017008655.1 | XP_016864144.1 | 6, 7 | 950 | |||
| XM_017008656.1 | XP_016864145.1 | 7, 8 | 1100 | |||
| XM_017008657.1 | XP_016864146.1 | 7, 8 | 1051 | |||
| XM_017008658.1 | XP_016864147.1 | 8, 9 | 1232 | |||
| XM_017008659.1 | XP_016864148.1 | 8, 9 | 1185 | |||
| XM_017008660.1 | XP_016864149.1 | 8, 9 | 1350 | |||
| GenBank mRNA | AB051499.1 | - | 7, 8 | 857 | ||
| AK094961.1 | - | 7, 8 | 972 | |||
| AK292484.1 | - | 7, 8 | 972 | |||
| AY426674.1 | - | 5, 6 | 686 | |||
| BC038667.1 | - | 7, 8 | 991 | 4823994 | ||

CEP44
Genomic Map