siRNA Details
Gene Symbol: TAS1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152232.2 | NP_689418.2 | 4, 5 | 1466 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
