siRNA Details
| Gene Symbol: TAS1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138697.3 | NP_619642.2 | 6 | 2529 | ||
| NM_177540.2 | NP_803884.1 | 5 | 1767 | |||
| XM_011542203.2 | XP_011540505.1 | 6 | 2895 | |||
| XM_011542206.2 | XP_011540508.1 | 5 | 2438 | |||
| XM_017002402.1 | XP_016857891.1 | 5 | 3081 | |||
| XM_017002403.1 | XP_016857892.1 | 4 | 2319 | |||
| GenBank mRNA | AF387618.1 | - | 5 | 2242 | ||
| AK292014.1 | - | 5 | 1736 | |||
| AK315805.1 | - | 6 | 2498 | |||
| BC136515.1 | - | 6 | 2529 | 9020505 | ||
| BC136516.1 | - | 6 | 2528 | 9020506 | ||
More Information
Set Membership: |
|

TAS1R1
Genomic Map