siRNA Details
| Gene Symbol: LNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305008.1 | NP_001291937.1 | 7 | 674 | ||
| NM_001305009.1 | NP_001291938.1 | 7 | 669 | |||
| NM_001305010.1 | NP_001291939.1 | 6 | 482 | |||
| NM_001305011.1 | NP_001291940.1 | 5 | 439 | |||
| NM_030650.2 | NP_085153.1 | 7 | 669 | |||
| NR_130941.1 | 8 | 958 | ||||
| XM_005246878.3 | XP_005246935.1 | 7 | 605 | |||
| XM_005246879.4 | XP_005246936.1 | 7 | 616 | |||
| XM_005246880.3 | XP_005246937.1 | 3 | 306 | |||
| XM_006712783.2 | XP_006712846.1 | 7 | 811 | |||
| XM_011511945.2 | XP_011510247.1 | 8 | 1020 | |||
| XM_017005055.1 | XP_016860544.1 | 3 | 214 | |||
| GenBank mRNA | AB051502.1 | - | 7 | 575 | ||
| AK056532.1 | - | 7 | 592 | |||
| AK301947.1 | - | 8 | 905 | |||
| AK302833.1 | - | 6 | 416 | |||
| AL832947.1 | - | 4 | 332 | |||
| BC105132.1 | - | 7 | 599 | 8144135 | ||
| BC105134.1 | - | 7 | 599 | 8144137 | ||
| BC110329.1 | - | 5 | 368 | 5732809 | ||
| BC143681.1 | - | 7 | 599 | 9052195 | ||
| BC143683.1 | - | 6 | 412 | |||
| CR936742.1 | - | 7 | 618 | |||

LNPK
Genomic Map