siRNA Details
| Gene Symbol: TUBB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030773.3 | NP_110400.1 | 3 | 465 | ||
| XM_017028085.1 | XP_016883574.1 | 3 | 659 | |||
| GenBank mRNA | AJ292757.1 | - | 3 | 465 | ||
| BC033679.1 | - | 3 | 298 | 5180793 | ||
| BI821931.1 | - | 3 | 298 | |||

TUBB1
Genomic Map