siRNA Details
| Gene Symbol: CLPTM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030782.4 | NP_110409.2 | 7 | 1067 | ||
| XM_011514144.2 | XP_011512446.1 | 7 | 1031 | |||
| GenBank mRNA | AB045223.1 | - | 7 | 824 | ||
| AK027306.1 | - | 7 | 881 | |||
| BC016399.1 | - | 5 | 453 | |||
| BC025305.1 | - | 7 | 885 | 3051368 | ||

CLPTM1L
Genomic Map