siRNA Details
| Gene Symbol: CLPTM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030782.4 | NP_110409.2 | 2 | 446 | ||
| XM_011514144.2 | XP_011512446.1 | 2 | 413 | |||
| GenBank mRNA | AB045223.1 | - | 2 | 203 | ||
| AK027306.1 | - | 2 | 260 | |||
| BC025305.1 | - | 2 | 264 | 3051368 | ||
| CD109529.1 | - | 2 | 474 | |||

CLPTM1L
Genomic Map