siRNA Details
| Gene Symbol: OR10W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207374.3 | NP_997257.2 | 1 | 1259 | ||
| GenBank mRNA | AY358251.1 | - | 1 | 1259 | ||
| BC132848.1 | - | 1 | 1158 | 40146870 | ||
| BC132850.1 | - | 1 | 1158 | 40146872 | ||

OR10W1
Genomic Map