siRNA Details
| Gene Symbol: RSPH6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030785.3 | NP_110412.1 | 3 | 1216 | ||
| XM_011527351.1 | XP_011525653.1 | 3 | 1192 | |||
| GenBank mRNA | AK093068.1 | - | 2 | 486 | ||
| AK098837.1 | - | 3 | 1161 | |||
| AK223347.1 | - | 3 | 1161 | |||
| AL136761.1 | - | 3 | 1188 | |||
| BC057785.1 | - | 3 | 1044 | |||
| BC136351.1 | - | 3 | 1097 | 9020338 | ||
| BC136352.1 | - | 3 | 1097 | 9020339 | ||

RSPH6A
Genomic Map